Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198777.1 | NP_001185706.1 | 4 | 527 | ||
NM_001198778.1 | NP_001185707.1 | 4 | 453 | |||
NM_001198779.1 | NP_001185708.1 | 4 | 566 | |||
NM_001324375.1 | NP_001311304.1 | 4 | 519 | |||
NM_001324376.1 | NP_001311305.1 | 4 | 566 | |||
NM_003591.3 | NP_003582.2 | 4 | 519 | |||
XM_011519743.1 | XP_011518045.1 | 6 | 681 | |||
XM_011519744.1 | XP_011518046.1 | 5 | 563 | |||
XM_011519745.1 | XP_011518047.1 | 5 | 615 | |||
XM_011519747.1 | XP_011518049.1 | 5 | 600 | |||
GenBank mRNA | AF126404.1 | - | 3 | 292 | ||
AK095217.1 | - | 4 | 527 | |||
AK294080.1 | - | 4 | 467 | |||
AK300491.1 | - | 4 | 411 | |||
AK310225.1 | - | 4 | 477 | |||
AL832733.1 | - | 4 | 503 | |||
BC009591.1 | - | 4 | 502 | 4104375 | ||
BC110901.1 | - | 4 | 507 | 6161736 | ||
DC401811.1 | - | 4 | 454 | |||
DC401936.1 | - | 4 | 519 | |||
U83410.1 | - | 4 | 438 |