Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146029.2 | NP_001139501.1 | 5 | 640 | ||
NM_001146030.2 | NP_001139502.1 | 6 | 578 | |||
NM_003612.4 | NP_003603.1 | 6 | 682 | |||
GenBank mRNA | AF030698.1 | - | 6 | 653 | ||
AF069493.1 | - | 6 | 658 | |||
AF071542.1 | - | 6 | 641 | |||
AK293280.1 | - | 5 | 636 | |||
AK312257.1 | - | 6 | 681 | |||
AM180445.1 | - | 6 | 641 | |||
AM180446.1 | - | 6 | 641 | |||
AM180447.1 | - | 6 | 641 | |||
AM180448.1 | - | 6 | 641 | |||
AM180449.1 | - | 6 | 641 | |||
AM180450.1 | - | 6 | 641 | |||
AM180451.1 | - | 6 | 641 | |||
BC101643.1 | - | 6 | 673 | 8069149 | ||
BC101647.1 | - | 6 | 673 | 8069153 | ||
DA136330.1 | - | 6 | 534 |