Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP4K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270425.1 | NP_001257354.1 | 4, 5 | 630 | ||
NM_003618.3 | NP_003609.2 | 4, 5 | 630 | |||
GenBank mRNA | AK291467.1 | - | 4, 5 | 596 | ||
AK314487.1 | - | 4, 5 | 467 | |||
BC071579.1 | - | 4, 5 | 622 | 5296205 |