Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003630.2 | NP_003621.1 | 2 | 424 | ||
GenBank mRNA | AB035307.1 | - | 2 | 308 | ||
AJ001625.1 | - | 2 | 225 | |||
AJ131389.1 | - | 2 | 221 | |||
AK025644.1 | - | 290 | ||||
AK300907.1 | - | 2 | 290 | |||
AY277600.1 | - | 2 | 181 | |||
BC014551.1 | - | 2 | 362 | 4333011 | ||
BC015506.1 | - | 2 | 323 | 3914956 | ||
BP377713.1 | - | 2 | 424 | |||
CR542062.1 | - | 2 | 162 |