Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPSNAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202502.1 | NP_001189431.1 | 10 | 1567 | ||
NM_003634.3 | NP_003625.2 | 10 | 1868 | |||
GenBank mRNA | AJ001258.1 | - | 10 | 1867 | ||
AK098367.1 | - | 10 | 1622 | |||
BC002371.2 | - | 10 | 1618 | 2822369 | ||
BC006473.1 | - | 10 | 1618 | 2822369 | ||
BC018869.2 | - | 9 | 1510 |