Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003635.3 | NP_003626.1 | 14 | 3266 | ||
XM_005270255.3 | XP_005270312.1 | 14 | 3229 | |||
XM_005270256.3 | XP_005270313.1 | 11 | 1471 | |||
XM_011540310.2 | XP_011538612.1 | 13 | 3835 | |||
XM_017016857.1 | XP_016872346.1 | 14 | 3295 | |||
GenBank mRNA | AB208870.1 | - | 8 | 3697 | ||
AK293527.1 | - | 10 | 1323 | |||
AK298957.1 | - | 12 | 1557 | |||
AK300520.1 | - | 10 | 1469 | |||
AK301251.1 | - | 9 | 1148 | |||
AK303648.1 | - | 15 | 3064 | |||
AK310580.1 | - | 9 | 1627 | |||
BC035711.1 | - | 14 | 3019 | 5587622 | ||
BC110588.1 | - | 12 | 2498 | 40011898 | ||
BC110589.1 | - | 12 | 2498 | 40012703 | ||
U36601.1 | - | 12 | 2489 |