Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303040.1 | NP_001289969.1 | 3 | 423 | ||
NM_001303041.1 | NP_001289970.1 | 2 | 311 | |||
NM_003637.4 | NP_003628.2 | 6 | 740 | |||
XM_005277436.3 | XP_005277493.1 | 5 | 491 | |||
XM_011510083.2 | XP_011508385.1 | 2 | 171 | |||
XM_017002622.1 | XP_016858111.1 | 4 | 697 | |||
XM_017002623.1 | XP_016858112.1 | 4 | 697 | |||
XM_017002624.1 | XP_016858113.1 | 4 | 697 | |||
XM_017002625.1 | XP_016858114.1 | 4 | 697 | |||
XM_017002627.1 | XP_016858116.1 | 4 | 697 | |||
GenBank mRNA | AF074015.1 | - | 6 | 586 | ||
AF112345.1 | - | 6 | 641 | |||
AK304155.1 | - | 3 | 279 | |||
AK304183.1 | - | 5 | 570 | |||
AK314255.1 | - | 6 | 589 | |||
BC140831.1 | - | 2 | 186 | 9021699 | ||
BC144637.1 | - | 2 | 186 | 9053168 |