Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199125.1 | NP_001186054.1 | 8 | 966 | ||
NM_001316354.1 | NP_001303283.1 | 8 | 1100 | |||
NM_001316356.1 | NP_001303285.1 | 7 | 883 | |||
NM_001316357.1 | NP_001303286.1 | 7 | 963 | |||
NM_001316358.1 | NP_001303287.1 | 6 | 833 | |||
NM_003653.3 | NP_003644.2 | 8 | 1013 | |||
XM_005256837.4 | XP_005256894.1 | 8 | 1032 | |||
XM_005256840.3 | XP_005256897.1 | 8 | 1077 | |||
XM_005256842.4 | XP_005256899.1 | 7 | 945 | |||
XM_017025246.1 | XP_016880735.1 | 7 | 871 | |||
GenBank mRNA | AF031647.1 | - | 8 | 918 | ||
AF098109.1 | - | 8 | 938 | |||
AK297706.1 | - | 8 | 988 | |||
AK302304.1 | - | 8 | 966 | |||
AK312476.1 | - | 8 | 954 | |||
AK316400.1 | - | 8 | 1045 | |||
BC001891.1 | - | 8 | 987 | 3537198 |