Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278589.1 | NP_001265518.1 | 12 | 1974 | ||
NM_003932.4 | NP_003923.2 | 12 | 2004 | |||
GenBank mRNA | AF116650.1 | - | 1 | 318 | ||
AK130770.1 | - | 12 | 1637 | |||
AY513286.1 | - | 12 | 1602 | |||
AY826824.1 | - | 12 | 1613 | |||
AY826825.1 | - | 12 | 1613 | |||
BC052982.1 | - | 12 | 1626 | 6046993 | ||
U17714.1 | - | 12 | 1610 |