Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321541.1 | NP_001308470.1 | 4 | 1844 | ||
NM_001321542.1 | NP_001308471.1 | 3 | 1889 | |||
NM_003676.3 | NP_003667.1 | 3 | 1779 | |||
XM_017002648.1 | XP_016858137.1 | 3 | 1819 | |||
GenBank mRNA | AY423730.1 | - | 3 | 1727 | ||
BC000961.2 | - | 3 | 1761 | 3450936 |