Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002877.1 | NP_001002877.1 | 15 | 1504 | ||
NM_001002878.1 | NP_001002878.1 | 15 | 1635 | |||
NM_001002879.1 | NP_001002879.1 | 14 | 1519 | |||
NM_003678.4 | NP_003669.4 | 14 | 1388 | |||
XM_005261797.1 | XP_005261854.1 | 15 | 1434 | |||
XM_005261798.1 | XP_005261855.1 | 16 | 1550 | |||
XM_005261799.1 | XP_005261856.1 | 15 | 1577 | |||
XM_005261801.3 | XP_005261858.1 | 14 | 1494 | |||
XM_006724347.1 | XP_006724410.1 | 16 | 1664 | |||
XM_017029027.1 | XP_016884516.1 | 16 | 2036 | |||
GenBank mRNA | AB023200.1 | - | 14 | 1355 | ||
AK025385.1 | - | 14 | 1338 | |||
AK098709.1 | - | 11 | 942 | |||
AK122673.1 | - | 17 | 2130 | |||
AK225752.1 | - | 13 | 1121 | |||
BC003615.2 | - | 14 | 1328 | 2988049 | ||
CR456542.1 | - | 13 | 1322 |