Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131027.1 | NP_001124499.1 | 6 | 1245 | ||
NM_003687.3 | NP_003678.2 | 7 | 1363 | |||
XM_017010002.1 | XP_016865491.1 | 5 | 1116 | |||
GenBank mRNA | AF153882.1 | - | 7 | 1326 | ||
BF689609.1 | - | 1 | 201 |