Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003940.2 | NP_003931.2 | 2 | 267 | ||
XM_011513269.1 | XP_011511571.1 | 2 | 214 | |||
XM_017007425.1 | XP_016862914.1 | 2 | 111 | |||
GenBank mRNA | AK290338.1 | - | 2 | 214 | ||
AK302404.1 | - | 2 | 76 | |||
BC016146.1 | - | 2 | 220 | 3910925 | ||
HM138083.1 | - | 2 | 186 | |||
U75362.1 | - | 2 | 280 |