Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003941.3 | NP_003932.3 | 2, 3 | 597 | ||
GenBank mRNA | AK289795.1 | - | 2, 3 | 510 | ||
AM295156.1 | - | 2, 3 | 260 | |||
BC052955.1 | - | 2, 3 | 547 | 5264663 | ||
CF593336.1 | - | 2, 3 | 626 | |||
CR749438.1 | - | 3, 4 | 502 | |||
D88460.1 | - | 2, 3 | 514 |