Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178040.1 | NP_001171511.1 | 8, 9 | 1230 | ||
NM_004177.4 | NP_004168.1 | 8, 9 | 1230 | |||
XM_005274195.3 | XP_005274252.1 | 8, 9 | 912 | |||
XM_005274198.2 | XP_005274255.1 | 8, 9 | 912 | |||
XM_011545221.2 | XP_011543523.1 | 7, 8 | 812 | |||
XM_017018188.1 | XP_016873677.1 | 8, 9 | 674 | |||
XM_017018189.1 | XP_016873678.1 | 8, 9 | 674 | |||
XM_017018190.1 | XP_016873679.1 | 8, 9 | 674 | |||
XM_017018191.1 | XP_016873680.1 | 8, 9 | 674 | |||
XM_017018192.1 | XP_016873681.1 | 8, 9 | 674 | |||
XM_017018193.1 | XP_016873682.1 | 8, 9 | 674 | |||
XM_017018194.1 | XP_016873683.1 | 8, 9 | 674 | |||
GenBank mRNA | AJ002076.1 | - | 8, 9 | 683 | ||
AK125660.1 | - | 8, 9 | 1230 | |||
AK290251.1 | - | 8, 9 | 799 | |||
AK297419.1 | - | 8, 9 | 837 | |||
AK312909.1 | - | 8, 9 | 846 | |||
BC007405.2 | - | 8, 9 | 873 | 3010338 | ||
BC007429.2 | - | 8, 9 | 873 | 3010338 | ||
BT006666.1 | - | 8, 9 | 683 | |||
BX538158.1 | - | 9, 10 | 1165 | |||
U32315.1 | - | 8, 9 | 721 | |||
X90581.1 | - | 7, 8 | 653 |