Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286074.1 | NP_001273003.1 | 39 | 5892 | ||
NM_004606.4 | NP_004597.2 | 38 | 5886 | |||
NM_138923.3 | NP_620278.1 | 38 | 5823 | |||
XM_005262295.1 | XP_005262352.1 | 39 | 5922 | |||
XM_005262296.1 | XP_005262353.1 | 39 | 5919 | |||
XM_005262297.4 | XP_005262354.1 | 39 | 5944 | |||
XM_006724682.3 | XP_006724745.1 | 38 | 6866 | |||
GenBank mRNA | AB209316.1 | - | 24 | 3343 | ||
AB300418.1 | - | 39 | 5819 | |||
D90359.1 | - | 38 | 5798 | |||
M73444.1 | - | 9 | 1111 | |||
R66536.1 | - | 32 |