Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261401.1 | NP_001248330.1 | 5 | 1699 | ||
NM_004622.2 | NP_004613.1 | 6 | 1779 | |||
NR_048556.1 | 5 | 1685 | ||||
NR_048557.1 | 4 | 1588 | ||||
NR_048558.1 | 3 | 1472 | ||||
NR_048559.1 | 4 | 1696 | ||||
GenBank mRNA | AK091915.1 | - | 6 | 1762 | ||
X78627.1 | - | 6 | 1625 |