Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004628.4 | NP_004619.3 | 6 | 864 | ||
NR_027299.1 | 5 | 740 | ||||
XM_011534093.2 | XP_011532395.1 | 6 | 821 | |||
GenBank mRNA | AK222844.1 | - | 5 | 557 | ||
AK289761.1 | - | 6 | 793 | |||
AK295711.1 | - | 6 | 682 | |||
AK311039.1 | - | 5 | 669 | |||
BC016620.1 | - | 6 | 775 | 4509957 | ||
D21089.1 | - | 6 | 865 | |||
HM113488.1 | - | 6 | 764 | |||
KJ534962.1 | - | 6 | 760 | |||
KJ535085.1 | - | 5 | 636 | |||
X65024.1 | - | 5 | 600 |