Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004628.4 | NP_004619.3 | 10 | 2062 | ||
NR_027299.1 | 9 | 1938 | ||||
XM_011534093.2 | XP_011532395.1 | 10 | 2019 | |||
GenBank mRNA | AK222844.1 | - | 9 | 1755 | ||
AK289761.1 | - | 10 | 1991 | |||
AK295711.1 | - | 10 | 1880 | |||
AK311039.1 | - | 9 | 1868 | |||
BC016620.1 | - | 10 | 1973 | 4509957 | ||
D21089.1 | - | 10 | 2063 | |||
EU530523.1 | - | 2 | 515 | |||
EU530529.1 | - | 2 | 269 | |||
EU530532.1 | - | 2 | 418 | |||
EU530534.1 | - | 2 | 269 | |||
HM113488.1 | - | 10 | 1962 | |||
KJ534962.1 | - | 10 | 1958 | |||
X65024.1 | - | 9 | 1798 |