Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199849.1 | NP_001186778.1 | 4 | 297 | ||
NM_001199850.1 | NP_001186779.1 | 3 | 204 | |||
NM_004632.3 | NP_004623.1 | 3 | 201 | |||
NM_033657.2 | NP_387506.1 | 3 | 270 | |||
XM_005245480.2 | XP_005245537.1 | 3 | 269 | |||
XM_011509972.2 | XP_011508274.1 | 3 | 200 | |||
XM_017002289.1 | XP_016857778.1 | 3 | 269 | |||
XM_017002290.1 | XP_016857779.1 | 4 | 293 | |||
XM_017002291.1 | XP_016857780.1 | 4 | 289 | |||
XM_017002294.1 | XP_016857783.1 | 4 | 289 | |||
GenBank mRNA | AK225869.1 | - | 4 | 297 | ||
AK293117.1 | - | 3 | 202 | |||
AK302281.1 | - | 3 | 204 | |||
BC002358.1 | - | 2 | 229 | |||
BC107487.1 | - | 3 | 201 | 40032516 | ||
BC107488.1 | - | 3 | 270 | 40032512 | ||
BT019494.1 | - | 2 | 113 | |||
CR749790.1 | - | 3 | 281 | |||
U18321.1 | - | 3 | 282 | |||
X83544.1 | - | 3 | 186 |