Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DAP3 SIRNA Assays ›
Gene Symbol:
DAP3
Gene Name:
death associated protein 3
Gene Aliases:
DAP-3, MRP-S29, MRPS29, bMRP-10
Chromosome Location:
Chr.1: 155688104 - 155739010 on Build GRCh38
UniGene ID:
Hs.516746
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139418
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DAP3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199849.1
NP_001186778.1
11
1047
NM_001199850.1
NP_001186779.1
9
852
NM_001199851.1
NP_001186780.1
9
831
NM_004632.3
NP_004623.1
10
951
NM_033657.2
NP_387506.1
10
1020
XM_005245480.2
XP_005245537.1
9
917
XM_005245481.2
XP_005245538.1
9
896
XM_005245482.4
XP_005245539.1
8
731
XM_011509972.2
XP_011508274.1
9
869
XM_017002289.1
XP_016857778.1
9
938
XM_017002290.1
XP_016857779.1
10
962
XM_017002291.1
XP_016857780.1
10
937
XM_017002292.1
XP_016857781.1
10
916
XM_017002293.1
XP_016857782.1
9
835
XM_017002294.1
XP_016857783.1
9
856
XM_017002295.1
XP_016857784.1
9
835
XM_017002296.1
XP_016857785.1
8
815
XM_017002297.1
XP_016857786.1
8
794
GenBank mRNA
AK225869.1
-
11
1046
AK293117.1
-
10
952
AK298201.1
-
9
839
AK302281.1
-
9
852
BC002358.1
-
9
979
BC107487.1
-
10
951
40032516
BC107488.1
-
10
1020
40032512
BT019494.1
-
9
863
CR749790.1
-
10
1031
U18321.1
-
10
1032
X83544.1
-
10
936
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial translational elongation
mitochondrial translational termination
apoptotic signaling pathway
structural constituent of ribosome
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-4h6qx:80/100.66.76.46:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline