Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243925.1 | NP_001230854.1 | 11 | 1567 | ||
NM_001243926.1 | NP_001230855.1 | 13 | 2046 | |||
NM_004635.4 | NP_004626.1 | 11 | 1552 | |||
GenBank mRNA | BC001662.2 | - | 11 | 1513 | 2988489 | |
BC007591.1 | - | 11 | 1489 | 3345889 | ||
BC010407.2 | - | 11 | 1511 | 3833667 | ||
BC068497.1 | - | 13 | 2000 | |||
U09578.1 | - | 11 | 1494 |