Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243925.1 | NP_001230854.1 | 11 | 2490 | ||
NM_001243926.1 | NP_001230855.1 | 13 | 2969 | |||
NM_004635.4 | NP_004626.1 | 11 | 2475 | |||
GenBank mRNA | BC001662.2 | - | 11 | 2436 | 2988489 | |
BC007591.1 | - | 11 | 2412 | 3345889 | ||
BC010407.2 | - | 11 | 2434 | 3833667 | ||
BC068497.1 | - | 13 | 2923 | |||
U09578.1 | - | 11 | 2417 |