Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPARCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128310.2 | NP_001121782.1 | 12 | 2731 | ||
NM_001291976.1 | NP_001278905.1 | 12 | 2738 | |||
NM_001291977.1 | NP_001278906.1 | 9 | 2389 | |||
NM_004684.5 | NP_004675.3 | 11 | 2629 | |||
GenBank mRNA | AK298039.1 | - | 12 | 1902 | ||
AK300956.1 | - | 12 | 2038 | |||
AK304494.1 | - | 9 | 2258 | |||
AK316533.1 | - | 12 | 2328 | |||
AL833430.1 | - | 11 | 2367 | |||
BC033721.1 | - | 11 | 2404 | 4939390 | ||
BX647713.1 | - | 12 | 2280 | |||
X82157.1 | - | 11 | 2356 | |||
X86693.1 | - | 11 | 2481 |