Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079539.1 | NP_001073007.1 | 6 | 1490 | ||
NM_005080.3 | NP_005071.2 | 5 | 1516 | |||
GenBank mRNA | AB076383.1 | - | 5 | 1476 | ||
AB076384.1 | - | 6 | 1450 | |||
AK093842.1 | - | 2 | 1868 | |||
BC000938.1 | - | 5 | 1473 | 3449394 | ||
BC012841.1 | - | 5 | 1500 | 3856898 | ||
BC015709.1 | - | 5 | 1409 | |||
M31627.1 | - | 5 | 1489 |