Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135823.1 | NP_001129295.1 | 3 | 2317 | ||
NM_005086.4 | NP_005077.2 | 3 | 2500 | |||
XM_011520853.2 | XP_011519155.1 | 3 | 2250 | |||
XM_011520854.2 | XP_011519156.1 | 3 | 2424 | |||
XM_011520855.2 | XP_011519157.1 | 3 | 2166 | |||
GenBank mRNA | AK092925.1 | - | 3 | 2312 | ||
AL136756.1 | - | 3 | 2483 |