Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034996.2 | NP_001030168.1 | 4, 5 | 440 | ||
NM_003973.4 | NP_003964.3 | 4, 5 | 376 | |||
GenBank mRNA | AK222750.1 | - | 1 | 335 | ||
AK223111.1 | - | 4, 5 | 329 | |||
AK290674.1 | - | 4, 5 | 336 | |||
AK292045.1 | - | 4, 5 | 334 | |||
AK300873.1 | - | 4, 5 | 329 | |||
BC000606.2 | - | 4, 5 | 342 | 3346054 | ||
BC005134.2 | - | 3, 4 | 307 | 3928410 | ||
BC009294.2 | - | 4, 5 | 339 | 4123062 | ||
BC016379.1 | - | 1 | 333 | |||
BC019651.1 | - | 1 | 314 | 4454086 | ||
BC022805.1 | - | 1 | 304 | |||
BC029036.1 | - | 4, 5 | 428 | 5183728 | ||
BC071913.1 | - | 1 | 336 | 6671085 | ||
BG564298.1 | - | 4, 5 | 376 | |||
BG776586.1 | - | 4, 5 | 359 | |||
CD298951.1 | - | 4, 5 | 443 | |||
D87735.1 | - | 4, 5 | 324 | |||
DQ118667.1 | - | 3, 4 | 307 | |||
L10376.1 | - | 1 | 312 | |||
U16738.1 | - | 3, 4 | 313 | |||
U92982.1 | - | 3, 4 | 241 |