Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005646.3 | NP_005637.3 | 6 | 1322 | ||
XM_005273234.3 | XP_005273291.1 | 6 | 1322 | |||
XM_011544263.2 | XP_011542565.1 | 6 | 1389 | |||
XM_017002194.1 | XP_016857683.1 | 6 | 1392 | |||
GenBank mRNA | U38847.1 | - | 6 | 1322 |