Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005646.3 | NP_005637.3 | 8 | 1620 | ||
XM_005273234.3 | XP_005273291.1 | 8 | 1620 | |||
XM_011544263.2 | XP_011542565.1 | 8 | 1687 | |||
XM_017002194.1 | XP_016857683.1 | 8 | 1690 | |||
XM_017002195.1 | XP_016857684.1 | 5 | 493 | |||
XM_017002197.1 | XP_016857686.1 | 5 | 493 | |||
GenBank mRNA | U38847.1 | - | 8 | 1620 |