Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL1X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139466.1 | NP_001132938.1 | 18 | 4568 | ||
NM_001139467.1 | NP_001132939.1 | 17 | 4423 | |||
NM_001139468.1 | NP_001132940.1 | 18 | 4558 | |||
NM_005647.3 | NP_005638.1 | 18 | 4687 | |||
XM_011545571.2 | XP_011543873.1 | 17 | 4409 | |||
GenBank mRNA | AB209608.1 | - | 13 | 4641 | ||
BC020374.1 | - | 1 | 1406 | |||
BC032708.1 | - | 18 | 4533 | 5496758 | ||
Y12781.1 | - | 18 | 4849 |