Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF354A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324339.1 | NP_001311268.1 | 4 | 2223 | ||
NM_005649.2 | NP_005640.2 | 5 | 2307 | |||
XM_011534645.2 | XP_011532947.1 | 5 | 2814 | |||
XM_017009791.1 | XP_016865280.1 | 5 | 2212 | |||
GenBank mRNA | AF116030.1 | - | 5 | 2314 | ||
AK290711.1 | - | 5 | 2202 | |||
BC047105.1 | - | 5 | 2264 | 5312219 | ||
D89928.1 | - | 5 | 2261 | |||
EU794664.1 | - | 5 | 2307 |