Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135099.1 | NP_001128571.1 | 13 | 1582 | ||
NM_005656.3 | NP_005647.3 | 13 | 1544 | |||
XM_005261043.2 | XP_005261100.1 | 12 | 1424 | |||
XM_011529731.2 | XP_011528033.1 | 13 | 1639 | |||
XM_011529733.1 | XP_011528035.1 | 11 | 1243 | |||
GenBank mRNA | AF123453.1 | - | 11 | 1410 | ||
AF270487.1 | - | 13 | 1523 | |||
AF329454.1 | - | 13 | 1537 | |||
AK222784.1 | - | 12 | 1455 | |||
AK291813.1 | - | 13 | 1499 | |||
AK296860.1 | - | 13 | 1582 | |||
AK313338.1 | - | 14 | 1844 | |||
BC035623.1 | - | 13 | 1521 | |||
BC051839.1 | - | 13 | 1515 | 6199625 | ||
U75329.1 | - | 12 | 1466 |