Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076785.2 | NP_001070253.1 | 4 | 422 | ||
NM_003983.5 | NP_003974.3 | 3 | 335 | |||
XM_011523433.1 | XP_011521735.1 | 3 | 437 | |||
XM_011523434.1 | XP_011521736.1 | 3 | 502 | |||
XM_011523438.1 | XP_011521740.1 | 2 | 892 | |||
XM_011523439.1 | XP_011521741.1 | 3 | 1021 | |||
GenBank mRNA | AK310866.1 | - | 3 | 301 | ||
BC028216.1 | - | 3 | 291 | 5214340 | ||
BC113100.1 | - | 3 | 322 | |||
CR749291.1 | - | 3 | 321 | |||
CR749475.1 | - | 3 | 304 | |||
D87432.1 | - | 3 | 370 | |||
DB107640.1 | - | 3 | 335 |