Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005677.3 | NP_005668.2 | 17 | 1598 | ||
NM_080538.2 | NP_536799.1 | 17 | 1727 | |||
NM_080539.3 | NP_536800.2 | 16 | 1496 | |||
GenBank mRNA | AF057036.1 | - | 17 | 1598 | ||
AJ225895.1 | - | 17 | 1592 | |||
AK128401.1 | - | 17 | 1727 | |||
AK289688.1 | - | 17 | 1617 | |||
AY150334.1 | - | 17 | 1578 | |||
AY150335.1 | - | 17 | 1578 | |||
AY150336.1 | - | 17 | 1578 | |||
AY150337.1 | - | 17 | 1578 | |||
AY150338.1 | - | 17 | 1578 | |||
AY150339.1 | - | 17 | 1578 |