Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSNAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005999.2 | NP_005990.1 | 4, 5 | 521 | ||
GenBank mRNA | AK313068.1 | - | 4, 5 | 459 | ||
BC010376.1 | - | 4, 5 | 479 | 4283594 | ||
BC011797.1 | - | 4, 5 | 496 | 4138517 | ||
BX537610.1 | - | 2, 3 | 3324 | |||
X95073.1 | - | 4, 5 | 523 |
Gene Symbol: TSNAX-DISC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028394.1 | 4, 5 | 521 | |||
NR_028395.1 | 4, 5 | 521 | ||||
NR_028397.1 | 4, 5 | 521 | ||||
NR_028399.1 | 4, 5 | 521 |