Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102420.2 | NP_001095890.1 | 4 | 863 | ||
NM_001102421.2 | NP_001095891.1 | 3 | 726 | |||
NM_001278243.1 | NP_001265172.1 | 4 | 442 | |||
NM_001278244.1 | NP_001265173.1 | 3 | 305 | |||
NM_001278245.1 | NP_001265174.1 | 4 | 439 | |||
NM_006007.3 | NP_005998.1 | 3 | 723 | |||
GenBank mRNA | AA837106.1 | - | 3 | 202 | ||
AF062346.1 | - | 3 | 446 | |||
AF062347.1 | - | 3 | 301 | |||
AI379933.1 | - | 3 | 434 | |||
AK025782.1 | - | 4 | 326 | |||
AK290849.1 | - | 3 | 446 | |||
BC011018.1 | - | 3 | 562 | 4331933 | ||
BC027707.2 | - | 4 | 813 | 3945047 | ||
BC073131.1 | - | 3 | 227 | 6137219 | ||
DA290666.1 | - | 3 | 305 | |||
DA392231.1 | - | 4 | 370 | |||
DA914616.1 | - | 4 | 386 |