Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297605.1 | NP_001284534.1 | 7 | 975 | ||
NM_003820.3 | NP_003811.2 | 8 | 1118 | |||
XM_006711019.2 | XP_006711082.1 | 8 | 790 | |||
XM_011542383.2 | XP_011540685.1 | 5 | 1161 | |||
XM_017002718.1 | XP_016858207.1 | 5 | 1241 | |||
XM_017002720.1 | XP_016858209.1 | 8 | 1836 | |||
XM_017002721.1 | XP_016858210.1 | 9 | 1246 | |||
GenBank mRNA | AB208808.1 | - | 5 | 3745 | ||
AF153978.1 | - | 8 | 1063 | |||
AF373877.1 | - | 8 | 818 | |||
AF373878.1 | - | 8 | 818 | |||
AK124010.1 | - | 6 | 2017 | |||
AY358879.1 | - | 8 | 899 | |||
BC002794.2 | - | 8 | 1084 | 3614650 | ||
BC018882.2 | - | 8 | 1084 | |||
BC029848.1 | - | 6 | 1055 | 5178561 | ||
BX343044.2 | - | 7 | 759 | |||
CR456909.1 | - | 8 | 818 | |||
U70321.1 | - | 8 | 1111 | |||
U81232.1 | - | 8 | 1082 |