Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193511.1 | NP_001180440.1 | 14 | 3619 | ||
NM_006301.3 | NP_006292.3 | 15 | 3520 | |||
XM_005269138.3 | XP_005269195.1 | 14 | 3547 | |||
XM_006719588.3 | XP_006719651.1 | 14 | 3441 | |||
XM_011538725.2 | XP_011537027.1 | 14 | 4424 | |||
XM_017019956.1 | XP_016875445.1 | 15 | 3448 | |||
GenBank mRNA | AB209453.1 | - | 5 | 5604 | ||
BC050050.1 | - | 11 | 2500 | 4824512 |