Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPDZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261406.1 | NP_001248335.1 | 46 | 6565 | ||
NM_001261407.1 | NP_001248336.1 | 45 | 6478 | |||
NM_003829.4 | NP_003820.2 | 46 | 6577 | |||
XM_005251622.4 | XP_005251679.1 | 47 | 6613 | |||
XM_005251623.4 | XP_005251680.1 | 45 | 6403 | |||
XM_006716885.3 | XP_006716948.1 | 47 | 6662 | |||
XM_006716886.3 | XP_006716949.1 | 48 | 6821 | |||
XM_006716887.3 | XP_006716950.1 | 47 | 6765 | |||
XM_006716888.3 | XP_006716951.1 | 46 | 6526 | |||
XM_006716889.3 | XP_006716952.1 | 46 | 6517 | |||
XM_006716891.3 | XP_006716954.1 | 45 | 6430 | |||
XM_017015252.1 | XP_016870741.1 | 46 | 6529 | |||
XM_017015253.1 | XP_016870742.1 | 45 | 6415 | |||
XM_017015254.1 | XP_016870743.1 | 46 | 6614 | |||
XM_017015255.1 | XP_016870744.1 | 45 | 6462 | |||
XM_017015256.1 | XP_016870745.1 | 44 | 6316 | |||
XM_017015257.1 | XP_016870746.1 | 44 | 6303 | |||
GenBank mRNA | AB210041.1 | - | 46 | 6578 | ||
AF093419.1 | - | 46 | 6404 | |||
AJ001319.1 | - | 12 | 1591 | |||
AK058011.1 | - | 7 | 2004 | |||
AK074721.1 | - | 16 | 2066 | |||
AK091945.1 | - | 22 | 2942 | |||
AK098775.1 | - | 14 | 1842 | |||
AK307988.1 | - | 23 | 2982 | |||
CR936648.1 | - | 31 | 6552 | |||
CX782625.1 | - | 2 | 425 |