Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278942.1 | NP_001265871.1 | 5 | 2228 | ||
NM_006753.5 | NP_006744.2 | 5 | 2229 | |||
NR_103874.1 | 5 | 2176 | ||||
GenBank mRNA | BC003001.1 | - | 5 | 2156 | ||
BC006197.2 | - | 5 | 2156 | 2820773 | ||
CB158198.1 | - | 1 | 352 |