Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003830.3 | NP_003821.1 | 2 | 514 | ||
XM_011527438.2 | XP_011525740.1 | 2 | 468 | |||
XM_017027419.1 | XP_016882908.1 | 2 | 468 | |||
GenBank mRNA | AF170484.1 | - | 2 | 411 | ||
AK225753.1 | - | 2 | 435 | |||
BC029896.1 | - | 2 | 416 | 5178889 | ||
DC324223.1 | - | 2 | 514 | |||
U71383.1 | - | 2 | 517 |
Gene Symbol: SIGLEC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098612.1 | NP_001092082.1 | 2 | 495 | ||
XM_017026113.1 | XP_016881602.1 | 2 | 441 | |||
GenBank mRNA | AK298284.1 | - | 2 | 417 | ||
AY854038.1 | - | 2 | 416 | |||
DC428881.1 | - | 2 | 514 |