Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006768.4 | NP_006759.3 | 2, 3 | 452 | ||
XM_005253944.4 | XP_005254001.1 | 2, 3 | 552 | |||
XM_011538789.2 | XP_011537091.1 | 2, 3 | 371 | |||
GenBank mRNA | AB208878.1 | - | 2, 3 | 407 | ||
AF035620.1 | - | 2, 3 | 387 | |||
AK290379.1 | - | 2, 3 | 392 | |||
AK308279.1 | - | 2, 3 | 299 | |||
AY332222.1 | - | 2, 3 | 387 | |||
BC136698.1 | - | 2, 3 | 387 | 9020688 | ||
BC136699.1 | - | 2, 3 | 387 | 9020689 |