Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006948.4 | NP_008879.3 | 3 | 557 | ||
GenBank mRNA | AK223282.1 | - | 3 | 515 | ||
AK312396.1 | - | 3 | 512 | |||
BC036370.2 | - | 3 | 519 | 4837136 | ||
BI548162.1 | - | 4 | 620 | |||
U04735.1 | - | 3 | 524 |