Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006948.4 | NP_008879.3 | 4 | 673 | ||
GenBank mRNA | AK128540.1 | - | 4 | 455 | ||
AK223282.1 | - | 4 | 631 | |||
AK312396.1 | - | 4 | 628 | |||
BC036370.2 | - | 4 | 635 | 4837136 | ||
U04735.1 | - | 4 | 640 |