Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006948.4 | NP_008879.3 | 5 | 1480 | ||
GenBank mRNA | AK128540.1 | - | 5 | 1262 | ||
AK223282.1 | - | 5 | 1438 | |||
AK299004.1 | - | 4 | 1215 | |||
BC036370.2 | - | 5 | 1442 | 4837136 | ||
U04735.1 | - | 5 | 1447 |