Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305033.1 | NP_001291962.1 | 4 | 3188 | ||
NM_001305035.1 | NP_001291964.1 | 6 | 3318 | |||
NM_001305036.1 | NP_001291965.1 | 5 | 3246 | |||
NM_001305037.1 | NP_001291966.1 | 5 | 3173 | |||
NM_001305038.1 | NP_001291967.1 | 5 | 3265 | |||
NM_001305039.1 | NP_001291968.1 | 6 | 3323 | |||
NM_001305040.1 | NP_001291969.1 | 7 | 3399 | |||
NM_006955.2 | NP_008886.1 | 5 | 3241 | |||
GenBank mRNA | AJ491697.1 | - | 5 | 2948 |