Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077516.1 | NP_001070984.1 | 8 | 1863 | ||
NM_001288777.1 | NP_001275706.1 | 8 | 1579 | |||
NM_006979.2 | NP_008910.2 | 7 | 2166 | |||
GenBank mRNA | AK225282.1 | - | 7 | 2090 | ||
AK301126.1 | - | 9 | 1765 | |||
BC000645.2 | - | 8 | 1801 | 3345970 | ||
D82060.1 | - | 7 | 2042 |