Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007115.3 | NP_009046.2 | 2 | 207 | ||
GenBank mRNA | AF086484.1 | - | 2 | 59 | ||
AJ419936.1 | - | 2 | 132 | |||
AJ421518.1 | - | 2 | 132 | |||
BC030205.1 | - | 2 | 176 | 5208773 | ||
M31165.1 | - | 2 | 200 |