Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF41 SIRNA Assays ›
Gene Symbol:
ZNF41
Gene Name:
zinc finger protein 41
Gene Aliases:
MRX89
Chromosome Location:
Chr.X: 47444691 - 47483671 on Build GRCh38
UniGene ID:
Hs.496074
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139721
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF41
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324139.1
NP_001311068.1
4
2564
NM_001324140.1
NP_001311069.1
5
2854
NM_001324141.1
NP_001311070.1
4
2593
NM_001324142.1
NP_001311071.1
5
2870
NM_001324143.1
NP_001311072.1
4
2906
NM_001324144.1
NP_001311073.1
5
2944
NM_001324145.1
NP_001311074.1
4
2609
NM_001324147.1
NP_001311076.1
5
2915
NM_001324148.1
NP_001311077.1
5
2960
NM_001324149.1
NP_001311078.1
4
2633
NM_001324150.1
NP_001311079.1
6
3042
NM_001324151.1
NP_001311080.1
5
2984
NM_001324152.1
NP_001311081.1
5
3030
NM_001324153.1
NP_001311082.1
5
2894
NM_001324154.1
NP_001311083.1
4
2781
NM_001324155.1
NP_001311084.1
4
2805
NM_001324156.1
NP_001311085.1
4
2586
NM_001324157.1
NP_001311086.1
4
2571
NM_007130.3
NP_009061.1
5
3257
NM_153380.3
NP_700359.1
5
3130
XM_006724550.3
XP_006724613.1
5
2983
XM_006724555.3
XP_006724618.1
5
3278
XM_017029810.1
XP_016885299.1
5
3358
XM_017029811.1
XP_016885300.1
5
3419
XM_017029812.1
XP_016885301.1
5
3340
XM_017029813.1
XP_016885302.1
5
2889
XM_017029814.1
XP_016885303.1
5
3329
XM_017029815.1
XP_016885304.1
5
3378
XM_017029816.1
XP_016885305.1
5
2817
XM_017029817.1
XP_016885306.1
5
2777
GenBank mRNA
AB209520.1
-
1
2966
AK290021.1
-
5
2911
AK294858.1
-
4
2506
BC015023.1
-
5
3140
3908304
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0