Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007146.2 | NP_009077.2 | 4 | 1004 | ||
XM_005257643.1 | XP_005257700.1 | 4 | 1001 | |||
XM_005257644.4 | XP_005257701.1 | 5 | 1157 | |||
XM_017025017.1 | XP_016880506.1 | 5 | 1208 | |||
XM_017025018.1 | XP_016880507.1 | 5 | 1060 | |||
GenBank mRNA | BX648862.1 | - | 3 | 321 | ||
D28118.1 | - | 4 | 902 |